Skip to Content
MilliporeSigma

SY3H3D67724C

Synthonix Corporation

6-Bromo-2,3,4-trifluoroaniline

Synonym(s):

6-bromo-2,3,4-trifluoroaniline

Sign In to View Organizational & Contract Pricing.

Select a Size

5 G

$34.50

10 G

$69.00

25 G

$103.50

Vendor SKU: B41885-5G

$34.50


Ships in 10 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
226
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C6H3BrF3N/c7-2-1-3(8)4(9)5(10)6(2)11/h1H,11H2

Inchi Key

AKJNEERVRJOBPD-UHFFFAOYSA-N

Storage temp.

Refrigerator

Purity

98%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

White to yellow powder or crystals

Looking for similar products? Visit Product Comparison Guide

MSDS

https://synthonix.com/41885.html

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service