Sign Into View Organizational & Contract Pricing
Select a Size
1 G
$23.00
5 G
$28.75
10 G
$46.00
Vendor SKU: C42692-1G
$23.00
Ships in 10 days - from Aldrich Partner.
About This Item
Skip To
Iupac Name
2-chloro-6-fluorophenylboronic acid
Inchi Code
1S/C6H5BClFO2/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3,10-11H
InChI key
NXSZSZJWZVLHAY-UHFFFAOYSA-N
storage temp.
Room Temperature
Purity
98%
Country of Origin
US
Shipping Temperature
Room Temperature
Physical Form
Crystal - Powder / Colour: White - Very pale yellow
Product Link
https://synthonix.com/42692.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://synthonix.com/42692.html
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service