Documents

SY3H3D67C547

Synthonix Corporation

2,5-Dimethylbenzoxazole

CAS Number:
Molecular Weight:
147.18
MDL number:

Iupac Name

2,5-dimethyl-1,3-benzoxazole

Inchi Code

1S/C9H9NO/c1-6-3-4-9-8(5-6)10-7(2)11-9/h3-5H,1-2H3

InChI key

XVQGFGKAPKEUFT-UHFFFAOYSA-N

storage temp.

Refrigerator

Purity

98%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Clear liquid / Colour: Pale yellow - Yellow

Product Link

https://synthonix.com/51756.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

H227, H315, H319

Precautionary statements

MSDS

https://synthonix.com/51756.html

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.