All Photos(1)

SY3H3D67F60E

Synthonix Corporation

2-Methyl-5-nitrophenol

CAS Number:
Molecular Weight:
153.14
MDL number:

Iupac Name

2-methyl-5-nitrophenol

Inchi Code

1S/C7H7NO3/c1-5-2-3-6(8(10)11)4-7(5)9/h2-4,9H,1H3

InChI key

UMFDLIXUUJMPSI-UHFFFAOYSA-N

storage temp.

Refrigerator

Purity

98%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Crystal - Powder / Colour: Pale yellow - Reddish yellow

Product Link

https://synthonix.com/39397.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

H302, H315, H319, H335

Precautionary statements

MSDS

https://synthonix.com/39397.html

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service