MilliporeSigma
All Photos(1)

SY3H8D526A65

Synthonix Corporation

(2S)-2-[(2-aminoacetyl)amino]-3-methylbutanoic acid

CAS Number:
Molecular Weight:
174.2
MDL number:

Iupac Name

(2S)-2-[(aminoacetyl)amino]-3-methylbutanoic acid

Inchi Code

1S/C7H14N2O3/c1-4(2)6(7(11)12)9-5(10)3-8/h4,6H,3,8H2,1-2H3,(H,9,10)(H,11,12)/t6-/m0/s1

InChI key

STKYPAFSDFAEPH-LURJTMIESA-N

storage temp.

Refrigerator

Purity

95%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Solid

Product Link

https://synthonix.com/69008.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

H302, H315, H319, H332, H335

Precautionary statements

MSDS

https://synthonix.com/69008.html

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service