Skip to Content
MilliporeSigma

SY3H98B97C5F

Synthonix Corporation

Methyl 2-Amidinoacetate Hydrochloride

Synonym(s):

methyl 3-amino-3-iminopropanoate hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Product SY3H98B97C5F is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
152.58
MDL number:

Skip To

Inchi Code

1S/C4H8N2O2.ClH/c1-8-4(7)2-3(5)6;/h2H,5-6H2,1H3;1H

Inchi Key

LBVIIJSCVFAODU-UHFFFAOYSA-N

Storage temp.

Refrigerator

Purity

97%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Solid

MSDS

https://synthonix.com/74667.html

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library