Skip to Content
MilliporeSigma

SY3H9A74179C

Synthonix Corporation

(6-Tetrahydropyran-2-yloxyhexyl)magnesium bromide, 0.50 M in THF

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Product SY3H9A74179C is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
289.5
MDL number:

Skip To

Marketplace Support
Need help or have questions about this product?
Contact Seller

Iupac Name

(6-((tetrahydro-2H-pyran-2-yl)oxy)hexyl)magnesium bromide

Inchi Code

1S/C11H21O2.BrH.Mg/c1-2-3-4-6-9-12-11-8-5-7-10-13-11;;/h11H,1-10H2;1H;/q;;+1/p-1

Inchi Key

ZEROYKPCLQPNQY-UHFFFAOYSA-M

Storage temp.

Refrigerator

Purity

0.50

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Solution

Product Link

https://synthonix.com/88276.html

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library