Skip to Content
MilliporeSigma

SY3H9A7BF75C

Synthonix Corporation

1,3,3-Trimethylpiperazine dihydrochloride

Synonym(s):

1,3,3-trimethylpiperazine dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H18N2Cl2
Molecular Weight:
201.14
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H16N2.2ClH/c1-7(2)6-9(3)5-4-8-7;;/h8H,4-6H2,1-3H3;2*1H

Inchi Key

YFTRYOMGQYCFPH-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library