Skip to Content
MilliporeSigma

TA9545855688

Targetmol Chemicals Inc.

Guanosine

Synonym(s):

2-(2-amino-6-hydroxy-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

Sign In to View Organizational & Contract Pricing.

Select a Size

2 G

$30.00

Vendor SKU: T6523-2G

$30.00


Ships in 7 days - from Aldrich Partner.


About This Item

Linear Formula:
C10H13O5N5
CAS Number:
Molecular Weight:
283.24
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C10H13N5O5/c11-10-13-7-4(8(19)14-10)12-2-15(7)9-6(18)5(17)3(1-16)20-9/h2-3,5-6,9,16-18H,1H2,(H3,11,13,14,19)

Inchi Key

NYHBQMYGNKIUIF-UHFFFAOYSA-N

Shipping Temperature

4

Purity

99.93%

Storage temp.

-80/-20

Country of Origin

CN

Physical Form

solid


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service