Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TA9H11E416B0

Targetmol Chemicals Inc.

Xylitol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
152.15
Product TA9H11E416B0 is currently not available for purchase. Contact Technical Service

Iupac Name

pentitol

Inchi Code

1S/C5H12O5/c6-1-3(8)5(10)4(9)2-7/h3-10H,1-2H2

InChI key

HEBKCHPVOIAQTA-UHFFFAOYSA-N

storage temp.

4

Purity

99.96%

Country of Origin

CN

Shipping Temperature

Room temperature/blue ice

Physical Form

solid

Product Link

https://www.targetmol.com/compound/Xylitol

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library