Skip to Content
MilliporeSigma

TA9H303760BC

Targetmol Chemicals Inc.

Resibufogenin

Synonym(s):

5-(7-hydroxy-9a,11a-dimethylhexadecahydronaphtho[1',2':6,7]indeno[1,7a-b]oxiren-1-yl)-2H-pyran-2-one

Sign In to View Organizational & Contract Pricing.

Select a Size


Inchi Code

1S/C24H32O4/c1-22-9-7-16(25)11-15(22)4-5-18-17(22)8-10-23(2)19(12-20-24(18,23)28-20)14-3-6-21(26)27-13-14/h3,6,13,15-20,25H,4-5,7-12H2,1-2H3

Inchi Key

ATLJNLYIJOCWJE-UHFFFAOYSA-N

Storage temp.

-80/-20

Purity

99.17%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

MSDS

https://www.targetmol.com/compound/Resibufogenin


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library