Skip to Content
MilliporeSigma

TA9H93CFBF91

Targetmol Chemicals Inc.

Ertapenem sodium

Synonym(s):

sodium 3-((2S,4S)-4-(((4R,5S,6S)-2-carboxy-6-((R)-1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-en-3-yl)thio)pyrrolidine-2-carboxamido)benzoate

Sign In to View Organizational & Contract Pricing.

Select a Size


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C22H25N3O7S.Na/c1-9-16-15(10(2)26)20(28)25(16)17(22(31)32)18(9)33-13-7-14(23-8-13)19(27)24-12-5-3-4-11(6-12)21(29)30;/h3-6,9-10,13-16,23,26H,7-8H2,1-2H3,(H,24,27)(H,29,30)(H,31,32);/q;+1/p-1/t9-,10-,13+,14+,15-,16-;/m1./s1

Inchi Key

ZXNAQFZBWUNWJM-HRXMHBOMSA-M

Storage temp.

-80/-20

Purity

98.44%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Looking for similar products? Visit Product Comparison Guide

MSDS

https://www.targetmol.com/compound/Ertapenem sodium


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service