MilliporeSigma
All Photos(1)

TA9H97BAEC65

Targetmol Chemicals Inc.

MZ 1

CAS Number:
Molecular Weight:
1002.66

Inchi Code

1S/C49H60ClN9O8S2/c1-28-30(3)69-48-41(28)42(33-12-14-35(50)15-13-33)54-37(45-57-56-31(4)59(45)48)23-39(61)51-16-17-65-18-19-66-20-21-67-26-40(62)55-44(49(5,6)7)47(64)58-25-36(60)22-38(58)46(63)52-24-32-8-10-34(11-9-32)43-29(2)53-27-68-43/h8-15,27,36-38,44,60H,16-26H2,1-7H3,(H,51,61)(H,52,63)(H,55,62)

InChI key

PTAMRJLIOCHJMQ-UHFFFAOYSA-N

Purity

99.31%

Country of Origin

US

Product Link

www.targetmol.com/compound/MZ 1

MSDS

www.targetmol.com/compound/MZ 1

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service