Skip to Content
MilliporeSigma

TA9H9A6681E3

Targetmol Chemicals Inc.

Inosine

Synonym(s):

(2S,5S)-2-(6-hydroxy-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C10H12O5N4
CAS Number:
Molecular Weight:
268.23
MDL number:

Inchi Code

1S/C10H12N4O5/c15-1-4-6(16)7(17)10(19-4)14-3-13-5-8(14)11-2-12-9(5)18/h2-4,6-7,10,15-17H,1H2,(H,11,12,18)/t4-,6?,7?,10-/m0/s1

Inchi Key

UGQMRVRMYYASKQ-AMKBJPJNSA-N

Shipping Temperature

4

Purity

99.89%

Storage temp.

-80/-20

Country of Origin

CN

Physical Form

solid

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library