Skip to Content
MilliporeSigma

TA9H9A9A447C

Targetmol Chemicals Inc.

Nourseothricin sulfate

Synonym(s):

(S)-3,6-diamino-N-((S)-4-amino-6-(((2R,3R,4S,5R,6R)-2-(((3aS,7R,7aS)-4,7-dihydroxy-3a,6,7,7a-tetrahydro-3H-imidazo[4,5-c]pyridin-2-yl)amino)-4-hydroxy-5-(hydroxy(imino)methoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)imino)-6-hydroxyhexyl)hexanimidic acid--sulfuric acid (1/1)

Sign In to View Organizational & Contract Pricing.

Select a Size

Product TA9H9A9A447C is currently not available for purchase. Contact Technical Service

Skip To

Inchi Code

1S/C25H46N10O9.H2O4S/c26-5-1-3-11(27)7-15(38)30-6-2-4-12(28)8-16(39)32-19-20(40)21(44-24(29)42)14(10-36)43-23(19)35-25-33-17-13(37)9-31-22(41)18(17)34-25;1-5(2,3)4/h11-14,17-21,23,36-37,40H,1-10,26-28H2,(H2,29,42)(H,30,38)(H,31,41)(H,32,39)(H2,33,34,35);(H2,1,2,3,4)/t11-,12-,13+,14+,17+,18-,19+,20-,21-,23+;/m0./s1

Inchi Key

PXUKRRCWPLLSJP-MTNJQIIPSA-N

Storage temp.

-80/-20

Purity

100%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

MSDS

https://www.targetmol.com/compound/Nourseothricin sulfate


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library