TA9H9A9A73C9
Targetmol Chemicals Inc.
N-Desmethylclozapine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
Molecular Weight:
312.8
Iupac Name
8-chloro-11-(piperazin-1-yl)-10H-dibenzo[b,e][1,4]diazepine
Inchi Code
1S/C17H17ClN4/c18-12-5-6-15-16(11-12)21-17(22-9-7-19-8-10-22)13-3-1-2-4-14(13)20-15/h1-6,11,19,21H,7-10H2
InChI key
HESZUPIXRNZIOI-UHFFFAOYSA-N
storage temp.
4
Purity
98.45%
Country of Origin
CN
Shipping Temperature
Room temperature/blue ice
Physical Form
solid
Product Link
https://www.targetmol.com/compound/N-Desmethylclozapine
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.