Sign In to View Organizational & Contract Pricing
Select a Size
100 G
$29.00
Vendor SKU: T203704-100G
$29.00
Ships in 14 days - from Aldrich Partner.
Skip To
Iupac Name
disodium thiosulfate pentahydrate
Inchi Code
1S/2Na.H2O3S2.5H2O/c;;1-5(2,3)4;;;;;/h;;(H2,1,2,3,4);5*1H2/q2*+1;;;;;;/p-2
InChI key
PODWXQQNRWNDGD-UHFFFAOYSA-L
storage temp.
-80/-20
Country of Origin
CN
Shipping Temperature
4
Physical Form
solid
Product Link
https://www.targetmol.com/compound/sodium-thiosulfate
MSDS
https://www.targetmol.com/compound/sodium-thiosulfate
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service