All Photos(1)

TA9545855791

Target Molecule Corp.

AST-1306 TsOH

CAS Number:
Molecular Weight:
621.09

Iupac Name

N-(4-{3-chloro-4-[(3-fluorobenzyl)oxy]anilino}-6-quinazolinyl)acrylamide 4-methylbenzenesulfonate

Inchi Code

1S/C24H18ClFN4O2.C7H8O3S/c1-2-23(31)29-17-6-8-21-19(11-17)24(28-14-27-21)30-18-7-9-22(20(25)12-18)32-13-15-4-3-5-16(26)10-15;1-6-2-4-7(5-3-6)11(8,9)10/h2-12,14H,1,13H2,(H,29,31)(H,27,28,30);2-5H,1H3,(H,8,9,10)

InChI key

ZMUKJEHWLJBODV-UHFFFAOYSA-N

storage temp.

-20*C

Purity

99.52%

Country of Origin

US

Shipping Temperature

0*C-4*C

Physical Form

Powder

Product Link

www.targetmol.com/compound/AST-1306 TsOH

MSDS

www.targetmol.com/compound/AST-1306 TsOH

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service