Documents

TA9H30375DD4

Target Molecule Corp.

Talc

CAS Number:
Molecular Weight:
385.31

Inchi Code

1S/3Mg.4O2Si.4H2O.3H/c;;;4*1-3-2;;;;;;;/h;;;;;;;4*1H2;;;/q3*+1;;;;;;;;;;;/p-3

InChI key

YGDRRZHQKHTOPI-UHFFFAOYSA-K

storage temp.

-20*C

Purity

98.00%

Country of Origin

US

Shipping Temperature

0*C-4*C

Physical Form

Powder

Product Link

www.targetmol.com/compound/Talc

MSDS

www.targetmol.com/compound/Talc

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.