Documents

TA9H93ED6B89

Target Molecule Corp.

Terpineol

CAS Number:
Molecular Weight:
617.01

Inchi Code

1S/4C10H18O/c2*1-8-4-6-9(7-5-8)10(2,3)11;2*1-8(2)9-4-6-10(3,11)7-5-9/h4,9,11H,5-7H2,1-3H3;9,11H,1,4-7H2,2-3H3;11H,4-7H2,1-3H3;9,11H,1,4-7H2,2-3H3/t9-;;;/m1.../s1

InChI key

YHBMRMXDATWKHX-NRYJBHHQSA-N

storage temp.

-20*C

Purity

98.15%

Country of Origin

US

Shipping Temperature

0*C-4*C

Physical Form

Powder

Product Link

www.targetmol.com/compound/Terpineol

MSDS

www.targetmol.com/compound/Terpineol

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.