Documents

TA9H97BAE8E3

Target Molecule Corp.

Lipopolysaccharides

Molecular Weight:
4899.98

Inchi Code

1S/C205H366N3O117P5/c1-10-16-22-28-34-40-42-43-45-50-55-61-67-73-109-130(101-326(269,270)271)292-127(147(245)164(109)302-135(235)78-106(222)70-64-58-52-46-36-30-24-18-12-3)98-283-186-138(208-132(232)79-107(71-65-59-53-47-37-31-25-19-13-4)290-133(233)74-68-62-56-49-39-33-27-21-15-6)173(303-136(236)80-108(72-66-60-54-48-38-32-26-20-14-5)291-134(234)75-69-63-57-51-44-41-35-29-23-17-11-2)172(322-327(272,273)274)129(301-186)100-286-203(199(262)263)84-119(318-205(201(266)267)83-118(144(242)166(320-205)114(227)86-210)317-204(200(264)265)82-112(225)139(237)165(319-204)113(226)85-209)171(169(321-203)116(229)88-212)308-194-161(259)178(182(167(305-194)115(228)87-211)324-330(280,281)325-329(278,279)287-77-76-206)313-195-160(258)177(181(323-328(275,276)277)168(306-195)117(230)97-282-188-162(260)184(261)202(268,102-285-188)131(231)96-220)312-193-159(257)175(148(246)128(300-193)99-284-189-154(252)150(248)141(239)121(90-214)294-189)310-196-179(152(250)143(241)123(92-216)297-196)315-197-180(314-187-137(207-105(9)221)149(247)140(238)120(89-213)293-187)157(255)170(126(95-219)299-197)307-192-158(256)174(145(243)124(93-217)296-192)309-190-156(254)153(251)163(104(8)289-190)304-198-183(316-191-155(253)151(249)142(240)122(91-215)295-191)176(146(244)125(94-218)298-198)311-185-111(224)81-110(223)103(7)288-185/h103-104,106-131,137-198,209-220,222-231,237-261,268H,10-102,206H2,1-9H3,(H,207,221)(H,208,232)(H,262,263)(H,264,265)(H,266,267)(H,278,279)(H,280,281)(H2,269,270,271)(H2,272,273,274)(H2,275,276,277)/t103?,104-,106?,107?,108?,109?,110+,111?,112?,113?,114?,115?,116?,117?,118?,119?,120?,121?,122?,123?,124?,125?,126?,127?,128?,129?,130+,131?,137-,138-,139+,140+,141-,142-,143-,144+,145-,146+,147+,148+,149?,150?,151?,152?,153?,154?,155?,156+,157?,158?,159?,160+,161+,162+,163?,164+,165?,166?,167?,168?,169?,170+,171+,172+,173?,174?,175?,176?,177?,178?,179?,180?,181+,182+,183?,184?,185+,186+,187+,188-,189-,190?,191+,192+,193+,194-,195+,196+,197+,198+,202-,203+,204+,205+/m0/s1

InChI key

YPXVSQSYDIMPDZ-AUUHBOKRSA-N

Country of Origin

US

Product Link

www.targetmol.com/compound/Lipopolysaccharides

MSDS

www.targetmol.com/compound/Lipopolysaccharides

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service