Documents

TA9H97BAEEF6

Target Molecule Corp.

Epidermal Growth Factor

CAS Number:
Molecular Weight:
6222.06

Inchi Code

1S/C270H401N73O83S7/c1-24-134(19)217(262(420)293-112-201(355)298-161(69-74-205(359)360)229(387)301-160(45-35-82-286-269(279)280)228(386)333-192(119-428)255(413)307-162(68-73-198(274)352)230(388)316-176(94-141-54-64-150(350)65-55-141)241(399)304-159(44-34-81-285-268(277)278)227(385)325-186(105-212(373)374)248(406)313-169(87-127(5)6)235(393)302-158(43-31-33-80-272)226(384)318-179(97-144-108-289-156-41-29-27-39-153(144)156)243(401)319-178(96-143-107-288-155-40-28-26-38-152(143)155)242(400)305-164(71-76-207(363)364)231(389)312-170(88-128(7)8)236(394)310-167(267(425)426)46-36-83-287-270(281)282)341-250(408)174(92-139-50-60-148(348)61-51-139)300-203(357)113-292-261(419)214(131(13)14)340-264(422)216(133(17)18)339-259(417)195(122-431)335-246(404)182(101-200(276)354)322-257(415)191(118-427)332-220(378)137(22)297-234(392)175(93-140-52-62-149(349)63-53-140)315-225(383)157(42-30-32-79-271)303-247(405)185(104-211(371)372)326-237(395)168(86-126(3)4)311-219(377)136(21)296-224(382)163(70-75-206(361)362)309-265(423)218(135(20)25-2)342-251(409)177(95-142-56-66-151(351)67-57-142)317-233(391)166(78-85-433-23)308-256(414)194(121-430)336-263(421)215(132(15)16)338-204(358)114-291-223(381)184(103-210(369)370)323-244(402)180(98-145-109-283-124-294-145)320-238(396)171(89-129(9)10)314-258(416)193(120-429)334-240(398)173(91-138-48-58-147(347)59-49-138)299-202(356)111-290-222(380)183(102-209(367)368)324-245(403)181(99-146-110-284-125-295-146)321-254(412)190(117-346)330-239(397)172(90-130(11)12)328-260(418)197-47-37-84-343(197)266(424)196(123-432)337-232(390)165(72-77-208(365)366)306-252(410)189(116-345)331-249(407)187(106-213(375)376)327-253(411)188(115-344)329-221(379)154(273)100-199(275)353/h26-29,38-41,48-67,107-110,124-137,154,157-197,214-218,288-289,344-351,427-432H,24-25,30-37,42-47,68-106,111-123,271-273H2,1-23H3,(H2,274,352)(H2,275,353)(H2,276,354)(H,283,294)(H,284,295)(H,290,380)(H,291,381)(H,292,419)(H,293,420)(H,296,382)(H,297,392)(H,298,355)(H,299,356)(H,300,357)(H,301,387)(H,302,393)(H,303,405)(H,304,399)(H,305,400)(H,306,410)(H,307,413)(H,308,414)(H,309,423)(H,310,394)(H,311,377)(H,312,389)(H,313,406)(H,314,416)(H,315,383)(H,316,388)(H,317,391)(H,318,384)(H,319,401)(H,320,396)(H,321,412)(H,322,415)(H,323,402)(H,324,403)(H,325,385)(H,326,395)(H,327,411)(H,328,418)(H,329,379)(H,330,397)(H,331,407)(H,332,378)(H,333,386)(H,334,398)(H,335,404)(H,336,421)(H,337,390)(H,338,358)(H,339,417)(H,340,422)(H,341,408)(H,342,409)(H,359,360)(H,361,362)(H,363,364)(H,365,366)(H,367,368)(H,369,370)(H,371,372)(H,373,374)(H,375,376)(H,425,426)(H4,277,278,285)(H4,279,280,286)(H4,281,282,287)/t134-,135-,136-,137-,154-,157-,158-,159-,160-,161-,162-,163-,164-,165-,166-,167-,168-,169-,170-,171-,172-,173-,174-,175-,176-,177-,178-,179-,180-,181-,182-,183-,184-,185-,186-,187-,188-,189-,190-,191-,192-,193-,194-,195-,196-,197-,214-,215-,216-,217-,218-/m0/s1

InChI key

GVUGOAYIVIDWIO-UFWWTJHBSA-N

storage temp.

-20*C

Purity

100.00%

Country of Origin

US

Shipping Temperature

0*C-4*C

Physical Form

Powder

Product Link

www.targetmol.com/compound/Epidermal Growth Factor

MSDS

www.targetmol.com/compound/Epidermal Growth Factor

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service