All Photos(1)

TA9H97BAEF2B

Target Molecule Corp.

Exendin-4 acetate

CAS Number:
Molecular Weight:
4246.68

Iupac Name

(4S,7S,10S,13S,16S,19S,22S,25S,28S,31S,34S,37S,40S,43S,46S,49S,52S,55S,58S,61S,67S)-4-((8S,11S,14S,17S)-8-(2-amino-2-oxoethyl)-1-((2S)-2-((2S)-1-((2S)-1-(2-((2S)-1-((2S)-2-((2S)-2-((2S)-2-(1-amino-3-hydroxy-1-oxopropan-2-ylcarbamoyl)pyrrolidine-1-carbonyl)pyrrolidine-1-carbonyl)pyrrolidin-1-yl)-1-oxopropan-2-ylamino)-2-oxoethylamino)-3-hydroxy-1-oxopropan-2-ylamino)-3-hydroxy-1-oxopropan-2-ylcarbamoyl)pyrrolidin-1-yl)-11-(4-aminobutyl)-18-(1H-indol-3-yl)-14-isobutyl-1,4,7,10,13,16-hexaoxo-3,6,9,12,15-pentaazaoctadecan-17-ylcarbamoyl)-67-(2-((S)-2-amino-3-(1H-imidazol-4-yl)propanamido)acetamido)-37-(3-amino-3-oxopropyl)-40-(4-aminobutyl)-10,58-dibenzyl-7-sec-butyl-25,28,31-tris(2-carboxyethyl)-49-(carboxymethyl)-16-(3-guanidinopropyl)-55,61-bis((R)-1-hydroxyethyl)-43,52-bis(hydroxymethyl)-13,46-diisobutyl-19-isopropyl-22-methyl-34-(2-(methylthio)ethyl)-6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60,63,66-henicosaoxo-5,8,11,14,17,20,23,26,29,32,35,38,41,44,47,50,53,56,59,62,65-henicosaazaheptacontane-1,70-dioic acid compound with acetic acid (1:1)

Inchi Code

1S/C184H282N50O60S.C2H4O2/c1-16-94(10)147(178(289)213-114(52-58-144(257)258)163(274)218-121(73-101-77-195-105-39-24-23-38-103(101)105)168(279)215-116(68-90(2)3)165(276)205-107(41-26-28-61-186)158(269)219-122(75-134(189)243)154(265)198-79-135(244)196-83-139(248)231-63-30-43-129(231)175(286)225-127(87-238)174(285)223-125(85-236)155(266)200-80-136(245)202-96(12)181(292)233-65-32-45-131(233)183(294)234-66-33-46-132(234)182(293)232-64-31-44-130(232)176(287)222-124(84-235)150(190)261)229-170(281)119(71-99-34-19-17-20-35-99)217-166(277)117(69-91(4)5)214-159(270)108(42-29-62-194-184(191)192)212-177(288)146(93(8)9)228-151(262)95(11)203-156(267)111(49-55-141(251)252)208-161(272)112(50-56-142(253)254)209-162(273)113(51-57-143(255)256)210-164(275)115(59-67-295-15)211-160(271)110(47-53-133(188)242)207-157(268)106(40-25-27-60-185)206-172(283)126(86-237)224-167(278)118(70-92(6)7)216-169(280)123(76-145(259)260)220-173(284)128(88-239)226-180(291)149(98(14)241)230-171(282)120(72-100-36-21-18-22-37-100)221-179(290)148(97(13)240)227-138(247)82-199-153(264)109(48-54-140(249)250)204-137(246)81-197-152(263)104(187)74-102-78-193-89-201-102;1-2(3)4/h17-24,34-39,77-78,89-98,104,106-132,146-149,195,235-241H,16,25-33,40-76,79-88,185-187H2,1-15H3,(H2,188,242)(H2,189,243)(H2,190,261)(H,193,201)(H,196,244)(H,197,263)(H,198,265)(H,199,264)(H,200,266)(H,202,245)(H,203,267)(H,204,246)(H,205,276)(H,206,283)(H,207,268)(H,208,272)(H,209,273)(H,210,275)(H,211,271)(H,212,288)(H,213,289)(H,214,270)(H,215,279)(H,216,280)(H,217,277)(H,218,274)(H,219,269)(H,220,284)(H,221,290)(H,222,287)(H,223,285)(H,224,278)(H,225,286)(H,226,291)(H,227,247)(H,228,262)(H,229,281)(H,230,282)(H,249,250)(H,251,252)(H,253,254)(H,255,256)(H,257,258)(H,259,260)(H4,191,192,194);1H3,(H,3,4)/t94-,95-,96-,97+,98+,104-,106-,107-,108-,109-,110-,111-,112-,113-,114-,115-,116-,117-,118-,119-,120-,121-,122-,123-,124-,125-,126-,127-,128-,129-,130-,131-,132-,146-,147-,148-,149-;/m0./s1

InChI key

YEAXWAGAAYUUBX-JVTOQCAZSA-N

Country of Origin

US

Product Link

www.targetmol.com/compound/Exendin-4 acetate

MSDS

www.targetmol.com/compound/Exendin-4 acetate

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service