Documents

TA9H97BAF1C6

Target Molecule Corp.

Enfuvirtide acetate

CAS Number:
Molecular Weight:
4551.99

Inchi Code

1S/C204H301N51O64.C2H4O2/c1-20-102(15)166(253-195(310)137(75-100(11)12)239-200(315)150(93-258)251-190(305)143(82-112-90-215-95-219-112)248-203(318)167(103(16)21-2)254-196(311)138(76-101(13)14)240-201(316)151(94-259)252-204(319)168(105(18)260)255-197(312)139(221-106(19)261)78-108-45-47-113(262)48-46-108)202(317)233-131(58-68-164(280)281)178(293)228-130(57-67-163(278)279)182(297)250-149(92-257)198(313)232-125(52-62-155(210)266)179(294)245-145(84-157(212)268)191(306)229-124(51-61-154(209)265)175(290)224-122(49-59-152(207)263)173(288)226-126(53-63-159(270)271)176(291)222-120(43-31-33-69-205)172(287)244-144(83-156(211)267)192(307)231-127(54-64-160(272)273)177(292)225-123(50-60-153(208)264)174(289)227-128(55-65-161(274)275)180(295)235-134(72-97(5)6)185(300)237-133(71-96(3)4)184(299)230-129(56-66-162(276)277)181(296)236-135(73-98(7)8)187(302)247-147(86-165(282)283)194(309)223-121(44-32-34-70-206)171(286)241-140(79-109-87-216-117-40-28-25-37-114(109)117)183(298)220-104(17)170(285)249-148(91-256)199(314)238-136(74-99(9)10)186(301)242-142(81-111-89-218-119-42-30-27-39-116(111)119)189(304)246-146(85-158(213)269)193(308)243-141(80-110-88-217-118-41-29-26-38-115(110)118)188(303)234-132(169(214)284)77-107-35-23-22-24-36-107;1-2(3)4/h22-30,35-42,45-48,87-90,95-105,120-151,166-168,216-218,256-260,262H,20-21,31-34,43-44,49-86,91-94,205-206H2,1-19H3,(H2,207,263)(H2,208,264)(H2,209,265)(H2,210,266)(H2,211,267)(H2,212,268)(H2,213,269)(H2,214,284)(H,215,219)(H,220,298)(H,221,261)(H,222,291)(H,223,309)(H,224,290)(H,225,292)(H,226,288)(H,227,289)(H,228,293)(H,229,306)(H,230,299)(H,231,307)(H,232,313)(H,233,317)(H,234,303)(H,235,295)(H,236,296)(H,237,300)(H,238,314)(H,239,315)(H,240,316)(H,241,286)(H,242,301)(H,243,308)(H,244,287)(H,245,294)(H,246,304)(H,247,302)(H,248,318)(H,249,285)(H,250,297)(H,251,305)(H,252,319)(H,253,310)(H,254,311)(H,255,312)(H,270,271)(H,272,273)(H,274,275)(H,276,277)(H,278,279)(H,280,281)(H,282,283);1H3,(H,3,4)/t102-,103-,104-,105+,120-,121-,122-,123-,124-,125-,126-,127-,128-,129-,130-,131-,132-,133-,134-,135-,136-,137-,138-,139-,140-,141-,142-,143-,144-,145-,146-,147-,148-,149-,150-,151-,166-,167-,168-;/m0./s1

InChI key

OCSYZCHOCWAFRR-BQABOTKESA-N

Purity

99.34%

Country of Origin

US

Product Link

www.targetmol.com/compound/Enfuvirtide acetate

MSDS

www.targetmol.com/compound/Enfuvirtide acetate

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service