MilliporeSigma
BKCO-SG604-1-10-26A-111-141
The Baker Company
Baker Company SterilGARD e3 Class II Type A2 Biosafety Cabinet
6 FT, 8 inch opening, w/Channel Stand, UV Light, and canopy exhaust connection
Shipped & Sold By Baker Co
A Trusted Marketplace Seller
Synonym(s):
BSC, Biosafety Cabinet, Containment Cabinet, Cell Culture Hood, Laminar Flow Hood
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
The Baker Company
General description
DESCRIPTION 6 ft SG w channel stand, UV Light, and canopy exhaust connection
General Description
SterilGARD® e3 Class II, Type A2 Biological Safety Cabinet
The most reliable, comfortable, and safe A2 cabinet in the industry is NSF certified and comes with a channel stand, UV light, FlexAIR™ canopy exhaust connection, and 8-inch sash opening.
Maximum protection is achieved through six technologies working in concert: our exclusive momentum air curtain, high-velocity return air slots, aerodynamically designed airfoil, optimized downflow, and unique air bypass armrest.
Features:
  • 6 ft width
  • Sash Opening: 8-inch
  • Adjustable Channel Stand
  • UV Light
  • FlexAIR canopy exhaust connection
  • StediFLOW™ VFD motor controller
  • ReadySAFE™ mode uses 50-75% less energy during off hours
  • Momentum air curtain and high-velocity return air slots minimize turbulence
  • Double wall design contains contaminated air under negative pressure
  • High-velocity return air slots
  • Exclusive NSF-approved cable ports
  • One-piece work surface
  • UniPressure™ Preflow plenum to extend filter life
  • Air bypass padded armrest
  • Air Pressure Monitoring
  • Maintenance access: Electrical, lamp, blower and filters easily accessible from the front of the cabinet
  • Stainless steel air diffuser/filter protector
  • Non-glare work surface and LED lighting reduce eyestrain
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)