50680
TrippNT
TrippNT® Safety Glass Holder
White PVC with 3 Compartments and Magnet Mount
Synonym(s):
safety glasses holder, eye-wear holder
About This Item
material
PVC
weight
0.93 lb
manufacturer/tradename
TrippNT
Related Categories
General description
TrippNT's Safety Glass Holder with 3 Compartments and Magnet Mount stores safety glasses keeping them secures when not in use. Constructed from white PVC it is lightweight and can be placed on counter surfaces or quickly mounted with it's magnet backing. All TrippNT products are made in the USA and come with a 100% Total Replacement Lifetime Guarantee.
- Rectangular Bin
- Magnet Backing for Easy Mounting
- Stores Safety Glasses When not in Use, Preventing Scratches
- Finish: Smooth, no rough or sharp edges
- Material: 1/4” White PVC
- Product Weight: 0.93 lbs
- Ships from: Kansas City, MO, USA
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.