All Photos(1)

V000358

Sigma-Aldrich

3-Methyl-1-butanol

p.a., 99.8%

Synonym(s):
Isopentyl alcohol, Isoamyl alcohol
Linear Formula:
(CH3)2CHCH2CH2OH
CAS Number:
Molecular Weight:
88.15
Beilstein:
1718835
EC Number:
MDL number:
PubChem Substance ID:

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 20 °C)

grade

p.a.

assay

99.8%

autoignition temp.

644 °F

expl. lim.

1.2-9 %, 100 °F

refractive index

n20/D 1.406 (lit.)

bp

130 °C (lit.)

mp

−117 °C (lit.)

density

0.809 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCO

InChI

1S/C5H12O/c1-5(2)3-4-6/h5-6H,3-4H2,1-2H3

InChI key

PHTQWCKDNZKARW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK Germany

WGK 1

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup

Certificate of Analysis

Certificate of Origin

Maud Contrant et al.
Nucleic acids research, 42(12), 7981-7996 (2014-05-17)
Micro (mi)RNAs are small non-coding RNAs with key regulatory functions. Recent advances in the field allowed researchers to identify their targets. However, much less is known regarding the regulation of miRNAs themselves. The accumulation of these tiny regulators can be...
Jane Melville et al.
PloS one, 9(7), e101847-e101847 (2014-07-31)
Molecular phylogenetics is increasingly highlighting the prevalence of cryptic species, where morphologically similar organisms have long independent evolutionary histories. When such cryptic species are known to be declining in numbers and are at risk of extinction due to a range...
Luca Bianco et al.
PloS one, 9(10), e110377-e110377 (2014-10-11)
High-density SNP arrays for genome-wide assessment of allelic variation have made high resolution genetic characterization of crop germplasm feasible. A medium density array for apple, the IRSC 8K SNP array, has been successfully developed and used for screens of bi-parental...
Rwik Sen et al.
Nucleic acids research, 42(15), 9892-9907 (2014-08-13)
Rrd1p (resistance to rapamycin deletion 1) has been previously implicated in controlling transcription of rapamycin-regulated genes in response to rapamycin treatment. Intriguingly, we show here that Rrd1p associates with the coding sequence of a galactose-inducible and rapamycin non-responsive GAL1 gene...
Shantala Arundhati Hari Dass et al.
Molecular ecology, 23(24), 6114-6122 (2014-08-22)
Male rats (Rattus novergicus) infected with protozoan Toxoplasma gondii relinquish their innate aversion to the cat odours. This behavioural change is postulated to increase transmission of the parasite to its definitive felid hosts. Here, we show that the Toxoplasma gondii...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service