V001661

Sigma-Aldrich

Acrylic acid

p.a., 99%

Linear Formula:
CH2=CHCOOH
CAS Number:
Molecular Weight:
72.06
Beilstein/REAXYS Number:
635743
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

vapor density

2.5 (vs air)

vapor pressure

4 mmHg ( 20 °C)

grade

p.a.

assay

99%

autoignition temp.

744 °F

expl. lim.

13.7 %

bp

139 °C (lit.)

mp

13 °C (lit.)

density

1.051 g/mL at 25 °C (lit.)

SMILES string

OC(=O)C=C

InChI

1S/C3H4O2/c1-2-3(4)5/h2H,1H2,(H,4,5)

InChI key

NIXOWILDQLNWCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Target Organs

Respiratory system

Hazard Codes

C,N

Risk Statement

10-20/21/22-35-50

Safety Statement

26-36/37/39-45-61

RIDADR

UN 2218 3(8) / PGII

WGK Germany

WGK 2

Flash Point(F)

119.3 °F - closed cup

Flash Point(C)

48.5 °C - closed cup

Adrie J J Straathof et al.
Applied microbiology and biotechnology, 67(6), 727-734 (2005-03-01)
Acrylic acid might become an important target for fermentative production from sugars on bulk industrial scale, as an alternative to its current production from petrochemicals. Metabolic engineering approaches will be required to develop a host microorganism that may enable such...
Akash Bachhuka et al.
ACS applied materials & interfaces, 7(42), 23767-23775 (2015-10-13)
Surface modification has been identified as an important technique that could improve the response of the body to implanted medical devices. Collagen production by fibroblasts is known to play a vital role in wound healing and device fibrous encapsulation. However...
Xiuping Zhu et al.
Environmental science & technology, 48(12), 7157-7163 (2014-05-28)
Several technologies, including pressure-retarded osmosis (PRO), reverse electrodialysis (RED), and capacitive mixing (CapMix), are being developed to recover energy from salinity gradients. Here, we present a new approach to capture salinity gradient energy based on the expansion and contraction properties...
Yagahira E Castro-Sesquen et al.
PLoS neglected tropical diseases, 8(10), e3211-e3211 (2014-10-03)
Detection of congenital T. cruzi transmission is considered one of the pillars of control programs of Chagas disease. Congenital transmission accounts for 25% of new infections with an estimated 15,000 infected infants per year. Current programs to detect congenital Chagas...
H Greim et al.
Chemosphere, 31(2), 2637-2659 (1995-07-01)
BUA compiled the available data on toxicity and ecotoxicity for several acrylic and methacrylic acid esters and their corresponding acids. A comparison of these data revealed a qualitative similarity in the toxicological and ecotoxicological properties of the compounds considered. The...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.