Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900532

Sigma-Aldrich

Diphenyl carbonate

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5O)2CO
CAS Number:
Molecular Weight:
214.22
Beilstein/REAXYS Number:
1074863
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

bp

301-302 °C (lit.)

mp

79-82 °C (lit.)

SMILES string

O=C(Oc1ccccc1)Oc2ccccc2

InChI

1S/C13H10O3/c14-13(15-11-7-3-1-4-8-11)16-12-9-5-2-6-10-12/h1-10H

InChI key

ROORDVPLFPIABK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

334.4 °F - closed cup

flash_point_c

168 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service