MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
UV-Visible Solvents
Two amber bottles of Uvasol® isooctane for spectroscopy
Ultraviolet-visible (UV-Vis) spectroscopy is a widely used analytical technique for measuring the absorbance of light in the ultraviolet (200–400 nm) and visible (400–800 nm) regions of the electromagnetic spectrum. This method relies on the principle that different compounds absorb light at specific wavelengths, enabling their identification and quantification.
Selecting the right solvent is critical for accurate UV-Vis analysis. Ideal solvents have high transparency in the measured wavelength range, minimizing background absorption that could interfere with analyte detection.
Products
UV/Vis spectroscopy (54)
HPLC (8)
gas chromatography (GC) (1)
hybridization (1)
ACS reagent (8)
Puriss P.A. (2)
Anhydrous (1)
Puriss (1)
Reagent Grade (1)
99% (22)
98% (1)
95% (2)
Sort by Relevance
Showing 1-20 of 54
Change View
Showing 1-20 of 54
Acetone
179124
Acetone
ACS reagent, ≥99.5%
Ethyl alcohol, Pure
459828
Ethyl alcohol, Pure
≥99.5%, HPLC/spectrophotometric grade, 200 proof, suitable for UV/Vis spectroscopy and HPLC
Toluene
34866
Toluene
suitable for HPLC, 99.9%
Hexane
270504
Hexane
≥95%, suitable for UV/Vis spectroscopy and HPLC, suitable for EPA ACB B21-02, EPA OTM-45, and EPA ACB B23-05b
Acetonitrile
439134
Acetonitrile
≥99.9%, gradient grade, suitable for UV/Vis spectroscopy, HPLC
2-Ethoxyethanol
128082
2-Ethoxyethanol
ReagentPlus®, 99%
Acetone
8.22251
Acetone
for preparative purposes, EMPLURA®
Ethyl alcohol, Pure
493511
Ethyl alcohol, Pure
≥95.0%, ACS spectrophotometric grade, 190 proof, suitable for UV/Vis spectroscopy
Acetone
32201
Acetone
puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
1,2,4-Trichlorobenzene
256412
1,2,4-Trichlorobenzene
suitable for HPLC, ≥99%
Cyclohexane
34855
Cyclohexane
suitable for HPLC, ≥99.7%
n-Hexane
1.04368
Hexane, mixture of isomers
≥95% (GC), suitable for UV/Vis spectroscopy, EMPLURA®
1-Butanol
34867
1-Butanol
suitable for HPLC, ≥99.7%
Ethylene glycol
293237
Ethylene glycol
spectrophotometric grade, ≥99%
Methanol
154903
Methanol
≥99.9%, ACS spectrophotometric grade, suitable for UV/Vis spectroscopy
n-Hexane
1.04372
Hexane
≥99% (GC), suitable for UV/Vis spectroscopy, Uvasol®
Phenol
1.00206
Phenol
GR for analysis ACS,Reag. Ph Eur
Chloroform
650471
Chloroform
HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Chloroform
366919
Chloroform
ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Isooctane
1.04718
Isooctane
for spectroscopy Uvasol®
You have viewed 1-20 of 54 results
Uvasol® Solvents
Uvasol® solvents are high-purity, optically transparent solvents designed for UV-Vis and infrared (IR) spectroscopy. With the highest UV transmittance and lowest UV absorbance, they ensure precise and reliable analytical results while minimizing the risk of misinterpretation.
These solvents comply with Ph. Eur. (European Pharmacopoeia) and USP (United States Pharmacopeia) methods, meeting stringent UV transmittance and absorbance standards as defined by Reag. Ph Eur and ACS specifications. Their exceptional batch-to-batch consistency reduces the need for repeat analyses, saving both time and costs. Additionally, Uvasol® solvents eliminate interferences from UV, IR, and fluorescence contamination, further enhancing analytical accuracy and application security.
Beyond UV-Vis applications, Uvasol® solvents are also suitable for IR spectroscopy. Commonly used solvents in this range include acetone, acetonitrile, 1-butanol, 2-propanol, chloroform, cyclohexane, dichloromethane, dimethyl sulfoxide (DMSO), ethanol, methanol, ethyl acetate, and n-heptane.
Acetone
Acetone is a polar aprotic solvent known for its strong solvating power, making it effective in dissolving a wide range of organic and inorganic compounds. It is highly transparent in the UV-Visible spectrum, with minimal absorbance above 330 nm, ensuring accurate spectroscopic measurement for analytes that absorb at lower wavelengths.
Acetonitrile
Acetonitrile is a highly transparent polar aprotic solvent, making it ideal for UV-Visible spectroscopy. With minimal absorbance below 210 nm, it allows precise measurement of analytes in the lower UV range. Its excellent solvating properties and compatibility with a wide range of reagents make it a preferred choice for chromatography, spectroscopic analyses, and other analytical applications.
Cyclohexane
Cyclohexane is a non-polar solvent commonly used for analyzing hydrophobic substances. It exhibits excellent transparency in the UV-Visible range, with minimal absorbance below 200 nm, making it suitable for detecting analytes that absorb in the deep UV region. Its low reactivity and high purity further enhance its suitability for spectroscopic and chromatographic applications.
Dimethyl Sulfoxide (DMSO)
Dimethyl sulfoxide (DMSO) is a highly polar aprotic solvent known for its strong solvating power and excellent transparency in the UV-Visible range. It exhibits minimal absorbance below 300 nm, making it suitable for spectroscopic analyses that require low background interference. 
Ethanol
Ethanol is a polar solvent with excellent transparency in the UV-Visible range, exhibiting minimal absorbance above 210 nm. Its compatibility with a wide range of biological molecules makes it a preferred solvent in biochemical assays, spectroscopic analyses, and pharmaceutical applications. Ethanol’s low UV absorbance ensures accurate measurement of analytes, particularly in biological and pharmaceutical research.
Isooctane
Isooctane is a highly non-polar solvent with exceptional transparency in the UV-Visible range, exhibiting very high transmittance even below 200 nm. Its low absorbance at deep UV wavelengths makes it an excellent choice for spectroscopic analyses, ensuring accurate and reliable analytical results. Isooctane is commonly used in UV-Vis spectroscopy and chromatographic applications where minimal solvent interference is critical.
Tetrahydrofuran (THF)
Tetrahydrofuran (THF) is a polar aprotic solvent with excellent transparency in the UV-Visible range, exhibiting low absorbance below 240 nm. This property makes it ideal for spectroscopic analyses, allowing accurate measurement of analytes in the UV region with minimal solvent interference. THF’s low viscosity enhances its handling and promotes efficient mixing with solutes, ensuring uniform and reliable solutions for analytical applications.