Viscosity Reduction Platform: Viscosity-reducing excipients for protein formulations
For biopharmaceuticals, delivering effective therapies frequently requires formulating highly concentrated protein solutions. However, high viscosity can impact processability, obstruct subcutaneous administration and limit therapeutic potential. Our proprietary combinations of viscosity-reducing excipients are designed to tackle these challenges head-on, to enable efficient processing, formulation, and delivery of protein-based therapeutics. While relying on viscosity reduction using a single excipient can be ineffective, the SAFC® platform of viscosity-reducing agent combinations empowers you to achieve optimal protein concentrations without compromising protein stability. Join us in overcoming the viscosity barrier and enhancing patient compliance with solutions that truly make a difference.
Key benefits and features
Subcutaneous administration of protein-based therapeutics improves patient compliance and helps avoid hospitalization—our Viscosity Reduction Platform enables subQ formulations even at protein concentrations that are typically challenging.
Efficient viscosity reduction, stable proteins
Reduce viscosity efficiently and maintain protein stability using synergistic excipient combinations.
Platform flexibility and support
Select the ideal combination for your protein from our Viscosity Reduction Platform, supported by expert application know-how.
Well characterized safety profile
Prior parenteral in-human applications and toxicological assessment available upon request.
The Viscosity Reduction Platform: Enabling subcutaneous formulation of biologics
Our Viscosity Reduction Platform enables subcutaneous formulation of highly concentrated protein biologics, bringing together increased concentrations and improved downstream processing.
Our offering comprises:
- Proprietary new excipient combinations to enable your target concentration. Commercial use licenses are available.
- Commercial supply of excipients to support your clinical development and commercialization
- Application know-how to support the selection of the most promising solution for your protein of interest
The ideal excipient combination tailored to your protein’s needs
As every protein is different, it is important to identify the most promising approach to viscosity reduction. The Viscosity Reduction Platform comprises excipients from two functional groups: an amino acid component (L-arginine, L-phenylalanine, L-ornithine monohydrochloride*) and an anionic component (benzenesulfonic acid, pyridoxine hydrochloride). These two components can act in synergy, leading in many cases to a superior viscosity reduction compared to a single excipient, whilst maintaining protein stability.
*L-ornithine monohydrochloride for R&D purposes only.
Emprove® Program: Supporting your risk mitigation efforts
Our commitment to advancing pharmaceutical development is backed by the Emprove® Program, which provides comprehensive documentation to expedite your journey to market. For high-risk applications such as parenteral administration, our Emprove® Expert excipient portfolio is particularly well suited as it features specified low endotoxin and bioburden levels to support risk mitigation efforts.
Frequently asked questions
What is a viscosity reducing agent and how does it work in protein solutions?
A viscosity reducing agent is a substance that decreases the viscosity of a fluid. In protein solutions, these agents work by reducing intermolecular interactions between protein molecules. This is crucial for high-concentration formulations, where increased interactions may result in clusters, high viscosities or protein aggregation. By incorporating viscosity reducing agents, manufacturers can achieve smoother, more manageable solutions that facilitate easier handling and administration, particularly in subcutaneous injections.
What are the benefits of using combinations of viscosity-reducing agents for high-concentration protein therapeutics?
Using proprietary combinations of viscosity-reducing excipients that are part of the Viscosity Reduction Platform offers several benefits: reduced viscosity of the protein solution improves processability which supports more efficient downstream processing e.g., using tangential flow filtration (TFF) and allows for higher concentrations in the final formulation; specific combinations of viscosity-reducing agents help maintain protein stability even at high protein concentrations that are typically prone to aggregation; and lower viscosity solutions are easier to inject and reduce pain at the injection site supporting a better patient experience, more convenient administration options, and improved patient compliance.
Which viscosity reducing additives are recommended for lowering the viscosity of protein solutions?
Recommended viscosity-reducing additives include a range of excipients that have been shown to be effective in protein formulations and are part of our Viscosity Reduction Platform offering. Combining different functional groups, amino acids and anionic agents, is shown to achieve superior viscosity reduction and protein stability compared to the individual excipients. The best suited excipient combination needs to be chosen based on the specific protein and formulation requirements.
How do you choose the right viscosity-decreasing agent for your formulation?
Choosing the right viscosity-decreasing agent involves several considerations: understanding the specific protein characteristics, defining the desired viscosity and intended route of administration (including feasible viscosity levels), and testing procedures to evaluate suitable excipients and excipient combinations that achieve the required viscosity-reducing effect. As each protein and protein formulation is different, a screening is recommended.
How can viscosity-reducing agents impact protein stability?
When reducing intermolecular interactions between different protein molecules, many viscosity-reducing excipients may also impact intramolecular interactions within the protein molecule. As the therapeutic function of the protein is highly dependent on its structure, which in turn is highly dependent on the interactions within the protein chain, any change in those intramolecular bonds may lead to (partial) unfolding and loss of protein functionality. The combinations of viscosity reducing agents that are part of the Viscosity Reduction Platform were successfully assessed for their viscosity-reducing effect and their ability to maintain protein stability.
Related webinars
Learn how to push concentration limits while maintaining protein stability with the Viscosity Reduction Platform and optimized single-pass TFF.
Formulating high protein concentration therapeutics during tangential flow filtration (TFF) can be limited by high protein viscosity. We will introduce the benefit of excipient combinations to reduce viscosity in TFF as well as the impact of a suitable cassette design to overcome these limitations.
The Viscosity Reduction Platform - Enabling Subcutaneous (subQ) Delivery
Related articles
- Addressing Viscosity Challenges for Subcutaneous Injections
This overview provides strategies for reducing the viscosity of high concentration drug formulations to enable subcutaneous injections for greater patient convenience and adherence.
- Viscosity reduction in high concentration TFF
This overview describes how use of viscosity-reducing excipients can address viscosity challenges in high concentration tangential flow filtration (TFF) and improve process economics.
- Pharma’s Almanac: Enabling stable, high-viscosity injectable drug products with new excipient combinations
About current issues with viscosity and injectable drugs, why viscosity has become a bigger concern today than in the past, and the approach to overcoming key limitations
- Pharma's Almanac: Viscosity reduction with an excipient platform and feasibility testing service
About current viscosity formulation challenges, the application of the Viscosity Reduction Platform, and benefits of the Feasibility Testing Service
- The Medicine Maker: High concentration protein therapeutics and their viscosity
About our scientists’ work on high concentration protein therapeutics and their viscosity, and what the solutions could mean for patients and their medicines
Related documents
- User Guide: Experimental procedures to evaluate the Viscosity Reduction Platform
Review a principle procedure to prepare a low volume of a highly concentrated protein formulation to allow for viscosity measurements.
- White Paper: Viscosity-reducing excipients for protein formulation
Read about the effect of protein viscosity on injection force and the platform’s ability to balance viscosity reduction with protein stability
- White Paper: Viscosity-reducing excipients for improvement of filtration processes
Read about benefits of viscosity-reducing excipients on productivity and final concentrations.
New solution for high-concentration biologics: How to reduce protein viscosity
Protein viscosity is one of the major obstacles in preparing highly concentrated protein formulations suitable for subcutaneous injection. Tobias Rosenkranz, Head of Biomolecule Formulation R&D at MilliporeSigma explains how the Viscosity Reduction Platform helps to solve formulation and processing challenges for more convenient administration.