Product Name
Ethylmalonic acid 97%
Product Number
102687
Product Brand
ALDRICH
CAS Number
601-75-2
Molecular Weight
132.11
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystal or Chunk(s)
TITRATION (T) NAOH 0.1M96.5 - 103.5 %
PURITY (GC AREA %)≥ 96.5 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE