Product Name
2,3,4,5,6-Pentafluorobenzaldehyde 98%
Product Number
103748
Product Brand
ALDRICH
CAS Number
653-37-2
Molecular Weight
196.07
Test
Specification
APPEARANCE (COLOR)Colorless to Yellow and Colorless to Green
APPEARANCE (FORM)Liquid or Solid
TITRATION97.5 - 102.5 %
TITRATION (METHOD)WITH HYDROXYLAMINE
PURITY (GC AREA %)≥ 97.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE