Product Name
2-Methoxybenzaldehyde 98%
Product Number
109622
Product Brand
ALDRICH
CAS Number
135-02-4
Molecular Weight
136.15
Test
Specification
APPEARANCE (COLOR)White to Light Yellow to Light Brown
APPEARANCE (FORM)Crystals or Solid or Liquid
TITRATION97.5 - 102.5 %
TITRATION (METHOD)WITH HYDROXYLAMINE
PURITY (GC AREA %)≥ 97.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE