Product Name
cis-Decahydronaphthalene 99%
Product Number
110469
Product Brand
ALDRICH
CAS Number
493-01-6
Molecular Weight
138.25
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE