Product Name
Vinylcyclopentane 99%
Product Number
111392
Product Brand
ALDRICH
CAS Number
3742-34-5
Molecular Weight
96.17
Test
Specification
APPEARANCECOLORLESS LIQUID
REFRACTIVE INDEX AT1.437 +/- 0.002
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
GAS LIQUID98.5% (MINIMUM)
 APPROVED MAY 16, 2007 RJM