Product Name
Hexylene glycol 99%
Product Number
112100
Product Brand
ALDRICH
CAS Number
107-41-5
Molecular Weight
118.17
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE