Product Name
4-Methoxybenzoic acid ReagentPlus®, 99%
Product Number
117390
Product Brand
ALDRICH
CAS Number
100-09-4
Molecular Weight
152.15
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Crystals or Powder with Chunk(s) or Crystals with Chunk(s)
TITRATION (T) NAOH 0.1M98.5 - 101.5 %
PURITY (HPLC AREA %)≥ 98.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE