Product Name
Phenylacetylene 98%
Product Number
117706
Product Brand
ALDRICH
CAS Number
536-74-3
Molecular Weight
102.13
Test
Specification
APPEARANCECOLORLESS TO DARK YELLOW LIQUID
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
GAS LIQUID97.5% (MINIMUM)
 REVISED JULY 22, 2008 RJM