Product Name
7-Hydroxy-4-methylcoumarin 97%
Product Number
128724
Product Brand
ALDRICH
CAS Number
90-33-5
Molecular Weight
194.18
Test
Specification
APPEARANCEWHITE TO OFF-WHITE POWDER, CRYSTALS, OR
 CRYSTALLINE POWDER
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
HIGH PRESSURE LIQUID96.5% (MINIMUM)
SOLUBILITY50 MG/ML, METHANOL; CLEAR, COLORLESS TO PALE
 YELLOW SOLUTION
 REVISED JUNE 29, 2005 RJM