Product Name
N-Benzylaniline 99%
Product Number
128937
Product Brand
ALDRICH
CAS Number
103-32-2
Molecular Weight
183.25
Test
Specification
APPEARANCECOLORLESS TO WHITE TO YELLOW TO AMBER POWDER
 CRYSTALS, CRYSTALLINE POWDER OR SOLID
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
TITRATION98.5%-101.5% (WITH HCLO4)
GAS LIQUID98.5 % (MINIMUM)
 APPROVED SEPTEMBER 29, 2003 JSB