Product Name
5-Nonanone 98%
Product Number
136948
Product Brand
ALDRICH
CAS Number
502-56-7
Molecular Weight
142.24
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 97.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE