Product Name
2-Methyl-3-nitrobenzoic acid 99%
Product Number
137847
Product Brand
ALDRICH
CAS Number
1975-50-4
Molecular Weight
181.15
Test
Specification
APPEARANCEWHITE TO ORANGE TO TAN POWDER, CRYSTALS,
 CRYSTALLINE POWDER AND/OR CHUNKS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
TITRATION98.5%-101.5% (WITH NAOH)
GAS LIQUID98.5% (MINIMUM)
 APPROVED DECEMBER 17, 2007 RJM