Product Name
Furil 98%
Product Number
138029
Product Brand
ALDRICH
CAS Number
492-94-4
Molecular Weight
190.15
Test
Specification
Appearance (Color)Yellow to Light Brown
Appearance (Form)Conforms to Requirements
 Powder or Crystals
Infrared spectrumConforms to Structure
Purity (GC)≥97.5 %