Product Name
1,3-Diphenylurea 98%
Product Number
142158
Product Brand
ALDRICH
CAS Number
102-07-8
Molecular Weight
212.25
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder
PURITY (HPLC AREA %)≥ 97.5 %
SOLUBILITY (COLOR)Colorless to Faint Yellow and Colorless to Faint Brown
SOLUBILITY (TURBIDITY)CLEAR TO VERY SLIGHTLY HAZY
SOLUBILITY (METHOD)50MG/ML, PYRIDIN
CARBON CONTENT71.7 - 75.4 %
NITROGEN CONTENT12.9 - 13.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE