Product Name
2-Cyclopenten-1-one ethylene ketal 95%
Product Number
147850
Product Brand
ALDRICH
CAS Number
695-56-7
Molecular Weight
126.15
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 94.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE