Product Name
Camphor 96%
Product Number
148075
Product Brand
ALDRICH
CAS Number
76-22-2
Molecular Weight
152.23
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Crystal or Chunk(s)
PURITY (GC AREA %)≥ 95.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE