Product Name
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride ≥97.0% (AT)
Product Number
15140
Product Brand
ALDRICH
CAS Number
68641-49-6
Molecular Weight
254.56
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
TITRATION≥ 97.0 %
TITRATION (METHOD)-
ASSAY (ELEMENTAL ANALYSIS)≥ 97.0 % (N)
SOLUBILITY (COLOR)Colorless to Faint Yellow
SOLUBILITY (TURBIDITY)Clear to Very Hazy
SOLUBILITY (METHOD)5% IN WATER
CARBON CONTENT28.31 % (THEORY)
HYDROGEN CONTENT3.17 % (THEORY)
NITROGEN CONTENT11.00 % (THEORY)
INFRARED SPECTRUMCONFORMS TO STRUCTURE
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
CARBON 13 NMR SPECTRUMCONFORMS TO STRUCTURE
PHOSPHORUS 31 NMR SPECTRUMCONFORMS TO STRUCTURE