Product Name
Boc-Gly-OH ≥99.0% (T)
Product Number
15420
Product Brand
ALDRICH
CAS Number
4530-20-5
Molecular Weight
175.18
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder or Crystals
TITRATION (T) NAOH 0.1M99.0 - 102.0 %
MELTING POINT86 - 89 C
RESIDUE ON IGNITION≤ 0.05 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE