Product Name
Boc-D-Phg-OH ≥99.0% (TLC)
Product Number
15487
Product Brand
ALDRICH
CAS Number
33125-05-2
Molecular Weight
251.28
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder
PURITY (TLC AREA %)≥ 99.0 %
SPECIFIC ROTATION (20/D)-141.0 ± 5.0 Degrees
CONCENTRATIONC=1 IN ETHANOL
CARBON CONTENT62.14 % (THEORY)
HYDROGEN CONTENT6.82 % (THEORY)
NITROGEN CONTENT5.57 % (THEORY)
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE